Learn More
Medchemexpress LLC Kojic acid dipalmita 50g
SDP
Supplier:  Medchemexpress LLC HYW09957950G
Encompass_Preferred
Kojic acid dipalmitate (Kojic dipalmitate) is a derivative of Kojic acid (HY-W050154) a fungal metabolite that can be produced by species of Aspergillus Acetobacter and Penicillium Kojic acid dipalmitate is a slow and reversible competitive inhibitor of tyrosinase Kojic acid dipalmitate can be used for skin lightening agent research[1]
Catalog No. 50-002-05878