Learn More
Med Vet International Kitty And Kanine Kollar, X-Small
Supplier:  Med Vet International JOR1085X
Encompass_Preferred
The extra small Petitie Kitty and Kanine Kollar was designed to be a sensible solution to the everyday patient management of an esophagostomy tube in cats and dogs. This collar has a neck size of 4-7in., is adjustable to fit securely around the animals neck, with a hole for the tube and a hook and loop strap to secure it. Circular cotton pads are supplied to place between the collar and the skin to protect the wound and absorb drainage from below. This collar for cats and dogs helps eliminate the need for owners to attempt to learn bandaging techniques, as well as unnecessary repeated visits to your veterinary office for re-bandaging. Features: Machine washable collar Ease of application and removal Lowered risk of tube removal Decreased risk of wound trauma Can be left in place for Radiographs, MRI, Chemotherapy, or other procedures Aesthetically pleasing for squeamish clients
Catalog No. 50-283-2247
 Restricted Item