Learn More
Sigma Aldrich Fine Chemicals Biosciences Kolliphor® EL, 61791-12-6, MFCD00130879, 500g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C5135500G
Encompass
pH-range 6.0 - 8.0, Synonym: Cremophor EL, Macrogolglycerol ricinoleate, PEG-35 castor oil, Polyoxyl 35 hydrogenated castor oil, Polyoxyl-35 castor oil; Kolliphor® EL is a nonionic solubilizer and emulsifier made by reacting castor oil with ethylene oxide. It is a pale yellow oily liquid that is clear at temperatures above 26�C. It has a faint but characteristic odour. Kolliphor® is a registered trademark of BASF SE.
Catalog No. 50-165-7076