Learn More
Selleck Chemical LLC KPT-276 1421919-75-6
SDP
Supplier:  Selleck Chemical LLC S725110MG
Encompass_Preferred
KPT-276 is an orally bioavailable selective CRM1 inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1918797