Learn More
Selleck Chemical LLC Selinexor (KPT-330) 5mg 1393477-72-9 ATG-010
SDP
Supplier:  Selleck Chemical LLC S72525MG
Encompass_Preferred
Selinexor (KPT-330, ATG-010) is an orally bioavailable selective CRM1 inhibitor. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0785745