Learn More
Medchemexpress LLC HY-100755 5mg Medchemexpress, KR-33493 CAS:1021497-97-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1007555MG
Encompass_Preferred
Medchemexpress, HY-100755 5mg KR-33493 CAS:1021497-97-1 KR-33493 is a potent inhibitor of Fas-mediated cell death (FAF1). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7778