Learn More
Sino Biological Recombinant Human KRAS Protein (G12C)
Supplier:  Sino Biological R0632DU20
Encompass_Preferred
Recombinant human untagged KRAS (G12C) (2-186) was expressed in E. coli cells. KRAS (G12C) is constitutively active.
Catalog No. 50-256-8011