Learn More
AURORA BIOLABS LLC Kras G12D, GppNHp loaded, 100 μg
Supplier:  AURORA BIOLABS LLC 57274123G GP100
Encompass
Kras is a member of the small G protein RAS family, which are a class of GTPases involved in cell signaling transduction. As an oncogene mutated in several different cancers, the G12V substitution mutation favors the activated, GTP-bound state of the protein.
Catalog No. NC3787935