Learn More
Medchemexpress LLC Kras G12d inhibitor 1 | 2621928-43-4 | 98.5% | C33H32F2N6O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13481110MG
Encompass_Preferred
KRAS G12D inhibitor 1 is a small-molecule research compound that selectively targets the KRAS G12D mutant. It is supplied as a solid or as a DMSO stock solution for in vitro biochemical and cellular studies and has been shown to inhibit KRAS G12D-mediated ERK phosphorylation with high potency.
  • Potent inhibition of KRAS G12D-mediated ERK phosphorylation (IC50 approximately 0.8 nM).
  • Suitable for biochemical and cellular assays investigating KRAS G12D-driven signaling.
  • Available as a solid and as a DMSO stock solution for flexible dosing.
  • Recommended storage conditions provided to preserve compound stability.
  • Reported high purity (approximately 98.5%) according to supplier documentation.
Catalog No. 50-002-61287