Learn More
Sino Biological Recombinant Human HRAS Protein (G12C, His & TEV Tag)
Supplier:  Sino Biological R0532CTH100
Encompass_Preferred
Recombinant human HRAS (G12C) (2-186) was expressed in E. coli cells using an N-terminal His-TEV tag. HRAS (G12C) is constitutively active.
Catalog No. 50-257-3371