Learn More
Medchemexpress LLC Kras4B protein, human (G12S, His) | >95.0% | 100UG

Supplier: Medchemexpress LLC HYP70153100UG
Recombinant human KRAS4B (G12S) with an N-terminal 6xHis tag, expressed in Escherichia coli and supplied as a lyophilized powder. The protein retains GDP/GTP binding and intrinsic GTPase activity and is intended for research applications investigating KRAS signaling and mutant-specific interactions. Supplied with high purity and recommended frozen storage to preserve activity.
- Contains G12S oncogenic mutation relevant for signaling studies.
- Includes N-terminal 6xHis tag for purification and detection.
- Expressed in Escherichia coli and provided as lyophilized powder.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Stable when stored at -20°C; aliquot to avoid freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.