Learn More
Medchemexpress LLC KRCA-0008 | 1472795-20-2 | MFCD28133395 | >95.0% | 609.12 g·mol^-1 | C30H37ClN8O4 | 10MG

Supplier: Medchemexpress LLC HY1233110MG
KRCA-0008 is a selective dual inhibitor of anaplastic lymphoma kinase (ALK) and activated Cdc42-associated kinase 1 (Ack1) used in cancer research. Reported biochemical IC50 values are 12 nM for ALK and 4 nM for Ack1. The compound is supplied as a powder for research use only.
- Selective dual ALK and Ack1 inhibitor
- Reported IC50: ALK 12 nM; Ack1 4 nM
- Molecular formula C30H37ClN8O4; molecular weight 609.12 g·mol^-1
- Supplied as a powder for laboratory use
- Recommended storage: powder at -20°C (long term) or 4°C (short term); in solution at -80°C
- Suitable for biochemical and cancer biology research (research use only)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.