Learn More
Medchemexpress LLC KRCA-0008 | 1472795-20-2 | MFCD28133395 | 96.2% | 609.12 g/mol | C30H37ClN8O4 | 50MG
SDP
Supplier:  Medchemexpress LLC HY1233150MG
Encompass_Preferred
KRCA-0008 is a potent, selective dual inhibitor of anaplastic lymphoma kinase (ALK) and activated Cdc42-associated kinase 1 (Ack1/TNK2), intended for research use in oncology and kinase signaling studies. It exhibits low-nanomolar biochemical potency and has shown in vivo activity in tumor models.
  • Potent dual ALK and Ack1 inhibition (ALK IC50 = 12 nM; Ack1 IC50 = 4 nM).
  • Suitable for cancer biology and kinase signaling research.
  • Reported molecular formula C30H37ClN8O4 and molecular weight 609.12 g/mol.
  • Available in multiple laboratory pack sizes in milligram quantities.
  • Purity reported by suppliers varies; consult the certificate of analysis for the supplied batch.
  • Research use only; not for human or veterinary application.
Catalog No. 50-002-39038