Learn More
KRISHGEN BIOSYSTEMS Immunoassay for the estimation of Faricimab in serum and plasma. Assay Range: 0.5 ng/ml – 10 ng/ml. Validated against ICH M10. Can be used for generics and biosimilars PK studies.
Supplier:  KRISHGEN BIOSYSTEMS KBI1326
Encompass_Preferred
Pack Size: 1 x 96 wells. Background: Faricimab is the first bispecific monoclonal antibody to target both vascular endothelial growth factor (VEGF) and angiopoietin 2 (Ang-2) inhibitor. A first in class treatment, Faricimab a humanized antibody that is composed of 2 heavy chains and 2 light chains, used for the treatment of neovascular age-related macular degeneration (nAMD) and diabetic macular edema (DME). It has been approved by the FDA and EMA in 2022. This Faricimab ELISA accurately quantifies faricimab in human samples. If you are looking to use animal samples, please contact us. Krishgen's range of bispecific ELISA for PK studies was Highly Commended in the CiteAb 2023 Innovation Award category. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Lot to Lot Consistency with Precision CV <15%.
Catalog No. NC2747092