Learn More
Medchemexpress LLC HY-12060 5mg Medchemexpress, KRN-633 CAS:286370-15-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY120605MG
Encompass_Preferred
Medchemexpress, HY-12060 5mg KRN-633 CAS:286370-15-8 KRN-633 is a potent VEGFR inhibitor with IC50s of 170, 160 and 125 nM for VEGFR1, VEGFR2 and VEGFR3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0945