Learn More
Bachem Osteoblast-Adhesive Peptide
SDP
Supplier:  Bachem 4029131.0025
Encompass_Preferred
Osteoblast-Adhesive Peptide, H-Lys-Arg-Ser-Arg-OH, 193613-75-1, C21H43N11O6, Mr 545.64, 25mg. KRSR selectively enhanced heparan sulfate-mediated osteoblast adhesion mechanisms. Elucidating such peptide sequence is of importance to the field of bone-cell/tissue engineering as well as to the design of proactive dental/orthopedic biomaterials. Note: Uses of this peptide are covered by U.S. Patent No. 6,262,017 B1 owned by Rensselaer Polytechnic Institute (RPI) located in Troy, New York, U.S.A. RPI authorizes purchasers of 5 grams or less per year to use this peptide for research purposes only. Other purchasers must have a written licence from RPI.
Catalog No. 50-194-6560