Learn More
Aviva Systems Biology Corp KRT3 Antibody - C-terminal region (ARP59767_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59767_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KRT3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human KRT3. Alternate names: CK3, FLJ95909, K3ApplicationWB. Species reactivity: Human, Rabbit
Catalog No. 50-286-818