Learn More
Medchemexpress LLC KS370G 10mg | 105955-01-9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11468310MG
Encompass_Preferred
KS370G (CAS 105955-01-9) is a small-molecule research compound reported to have oral hypoglycemic and cardiovascular protective activity. It acts on pathways including TGF-beta/Smad and is supplied as a solid for preclinical pharmacology and cellular studies.
  • Orally active hypoglycemic and cardiovascular protective agent.
  • High reported purity: 99.71%.
  • Molecular weight 283.32, formula C17H17NO3.
  • Supplied as a solid suitable for preclinical and in vitro assays.
  • Available in a 10 mg research sample size.
Catalog No. 50-002-29610