Learn More
Medchemexpress LLC KSC-34 | 2226201-97-2 | 99.9% | 429.95 g/mol | C21H28ClN7O | 100 MG

Supplier: Medchemexpress LLC HY117570100MG
KSC-34 is a small-molecule covalent modifier and selective inhibitor of protein disulfide isomerase A1 (PDIA1), used as a biochemical probe in in vitro and cell-based studies.
- Covalent modifier of protein disulfide isomerase A1 (PDIA1).
- Selective a-site inhibitor with reported IC50 ≈ 3.5 μM.
- Contains an alkyne functional group compatible with click chemistry applications.
- Molecular formula C21H28ClN7O, molecular weight 429.95 g/mol.
- Supplied as a research reagent for biochemical and cell-based assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.