Learn More
Medchemexpress LLC KSD 2405 | 620-24-6 | 99.2% | C7H8O2 | 10 G

Supplier: Medchemexpress LLC HY7844610G
KSD 2405 is an endogenous metabolite intended for research use only. It targets Endogenous Metabolite and its pathway is Metabolic Enzyme/Protease.
- Endogenous metabolite
- Molecular weight of 124.14
- Chemical formula: C7H8O2
- Appears as a solid with a light brown to brown color
- Soluble in DMSO (100 mg/mL)
- Powder can be stored at -20°C for 3 years or 4°C for 2 years
- When in solvent, store at -80°C for 2 years or -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.