Learn More
Selleck Chemical LLC Ksg hydrochloride hydrate
SDP
Supplier:  Selleck Chemical LLC S59685MG
Encompass_Preferred
Kasugamycin (Ksg) hydrochloride hydrate an antibiotic which binds both the 30S and 70S ribosome specifically inhibits translation initiation of canonical but not of leaderless messenger RNAs
Catalog No. 50-000-04098