Learn More
Medchemexpress LLC KT-333 | 2502186-79-8 | MFCD20760249 | 97.4% | 1257.78 g·mol⁻¹ | C60H74ClN10O14PS | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15673010MG
Encompass_Preferred
KT-333 is a small-molecule molecular glue degrader that selectively induces proteasomal degradation of STAT3 by recruiting the VHL E3 ubiquitin ligase. It is supplied as a solid for preclinical research and demonstrates selective STAT3 degradation and antitumor activity in hematologic malignancy models.
  • Selective STAT3 degradation via VHL-mediated ubiquitination.
  • Demonstrates antitumor activity in preclinical hematologic malignancy models.
  • High purity suitable for preclinical research (≈97.4%).
  • Supplied as a white to off-white solid; recommended storage as powder at -20°C.
  • Available in small research pack sizes, including 10 mg.
Catalog No. 50-002-87850