Learn More
TARGETMOL CHEMICALS INC KT-474 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T744111MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. KT-474 (KYM-001) is a PROTAC degrader targeting interleukin-1 receptor-associated kinase 4 (IRAK4) with antitumor activity for the study of autoimmune, inflammatory, and cardiovascular disease disorders.
Catalog No. 50-287-6225