Learn More
Cayman Chemical KT 5720, 108068-98-0, 50 µg
SDP
Supplier:  Cayman Chemical 1001101150
Encompass_Preferred
A potent inhibitor of PKA signaling that acts through competitive inhibition of ATP (Ki = 60 nM)
Catalog No. NC0265282
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.