Learn More
Medchemexpress LLC HY-114926 5mg Medchemexpress, KT185 CAS:1472640-86-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1149265MG
Encompass_Preferred
Medchemexpress, HY-114926 5mg KT185 CAS:1472640-86-0 KT185 is an orally-bioavailable, brain-penetrant and selective ABHD6 inhibitor, with an IC 50 0.21 nM in Neuro2A cells [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2500