Learn More
Hach Company Alkali Buffer for Brine (Pack of 4)
Supplier:  Hach Company 2298504
Encompass
This alkali buffer is used in the APA 6000 low range hardness analyzer. Pack of 4
Catalog No. 50-199-5752