Learn More
Hach Company Ascorbic Acid Reagent Package (2.9L) for S5000 Low Range Phosphate
Supplier:  Hach Company 2600303
Encompass
Ascorbic Acid reagent package (dilution solution and raw material) for the Series 5000 Low Range Phosphate process analyzer. Replenish every 30 days for continuous operation.
Catalog No. 50-199-6307