Learn More
Hach Company CEL Advanced Drinking Water Laboratory
Supplier:  Hach Company 251235
Encompass
The CEL Advanced Drinking Water Laboratory provides complete field capabilitities, with instrumentation and tests that cover every major drinking water parameter. Kit includes DR 900 Colorimeter, HQ40d Multi-meter, PHC201 pH probe (1 meter cable), and CDC401 conductivity probe (1 meter cable), as well as Digital Titrator, reagent sets, apparatus, illustrated instrument manual, procedure manual CD and sturdy field case.
Catalog No. 50-199-4787