Learn More
Hach Company Intellical™ ISENH418101 Ammonium (NH₄⁺) Ion Selective Electrode (ISE) with Calibration Reagents Pack
Supplier:  Hach Company ISENH4181AP
Encompass
The Intellical™ Ammonium ISE Analysis Package contains all you need to start measuring ammonium (NH4+) with your HQD meter instrument: The Intellical™ ISENH4181 Ammonium ISE with 1 m cable, 500 mL bottles of 10 mg/L and 100 mg/L of Hach nitrogen-ammonia standard solutions, pack of 100 Hach ammonium ionic strength adjustor (ISA) Powder Pillows.
Catalog No. 50-199-7732