Learn More
Hach Company HQ430D Laboratory Conductivity/TDS Meter Package with 4 Poles Graphite CDC401 Cell
Supplier:  Hach Company 8509600
Encompass
Designed for water quality experts, the Hach HQ430D Laboratory Multi-Meter is an advanced laboratory meter that takes the guesswork out of measurements. HQD meters connect with smart Intellical electrodes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced HQD Laboratory Meter is designed to withstand years of continuous lab use. HQD meters feature a simple user interface that does not require manuals or training to operate. The Intellical CDC401 with 1 m cable CDC401 is a digital, graphite, 4-pole conductivity electrode, ideal for measuring electrical conductivity, salinity, resistivity, or total dissolved solids (TDS) in wastewater, drinking water, or general water quality applications.
Catalog No. 50-199-7268