Learn More
Hach Company Cadmium Reagent Set, Dithizone (No Air Shipments)
Supplier:  Hach Company 2242200
Encompass
For determination of cadmium by the Dithizone method. Hach method 8017. Range: 0.7 - 80 μg/L Set includes reagents for 100 tests. For air shipments, please order 2242201.
Catalog No. 50-199-5608