Learn More
Hach Company Volatile Acids Reagent Set, Esterification
Supplier:  Hach Company 2244700
Encompass
For determination of Volatile Acids by the Esterification method. Hach Method 8196. Range: Up to 2800 mg/L as Acetic Acid. Set Includes reagents for 100 tests.
Catalog No. 50-199-5631