Learn More
Medchemexpress LLC KTX-582 | 2573298-13-0 | 98.7% | 872.93 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1482741MG
Encompass_Preferred
KTX-582 is a potent IRAK4 degrader with DC50 values of 4 nM and 5 nM for IRAK4 and Ikaros, respectively. It can induce apoptosis in MYD88MT DLBCL and is efficient at inducing in vivo tumor regressions in a lymphoma model.
  • Potent IRAK4 degrader
  • Induces apoptosis in MYD88MT DLBCL
  • Efficient at inducing in vivo tumor regressions in lymphoma models
  • Degrades IRAK4 in whole blood monocyte and lymphocyte with IC50s of <0.05 μM
  • Inhibits IRAK4 in human whole blood LPS TNFα with an IC50 of 0.05-1 μM
Catalog No. 50-003-62864