Learn More
Medchemexpress LLC Ktx-582 intermediate-1 | 845508-29-4 | 326.41 | C16H22O5S | 25 MG

Supplier: Medchemexpress LLC HY4943625MG
KTX-582 intermediate-1 is an organic synthesis intermediate used in the preparation of the IRAK4 degrader KTX-582. It is supplied for research use and is characterized by its chemical identifiers and formula.
- Used as an intermediate in small-molecule synthesis
- Cas number 845508-29-4
- Molecular formula C16H22O5S
- Molecular weight 326.41
- Intended for research use only
- Pack size commonly 25 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.