Learn More
Selleck Chemical LLC KU-0063794 5mg 938440-64-3
SDP
Supplier:  Selleck Chemical LLC S12265MG
Encompass_Preferred
KU-0063794 is a potent and highly specific dual-mTOR inhibitor of mTORC1 and mTORC2 with IC50 of ~10 nM in cell-free assays; no effect on PI3Ks. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-837-4