Learn More
Apexbio Technology LLC KU 55933 587871-26-9 50mg
SDP
Supplier:  Apexbio Technology LLC A460550
Encompass_Preferred
KU 55933 (CAS 587871-26-9) is a selective inhibitor of ATM kinase with an IC50 of 13 nM ATM kinase promotes insulin-induced Akt activation via phosphorylation of Akt at Ser473 KU 55933 inhibition of ATM significantly reduces Akt Ser473 phosphorylation in insulin- or IGF-I-treated MDA-MB-453 and PC-3 cells resulting in suppressed proliferation cell cycle arrest at G1 phase and decreased cyclin D1 levels KU 55933 serves as a tool for studying ATM-related signaling and Akt-driven cancer cell growth regulation
Catalog No. 50-000-61638