Learn More
Apexbio Technology LLC KU-60019, 50mg. Cas: 925701-49-1 MFCD: MFCD18384974. ATM kinase inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A833650
Encompass_Preferred
KU-60019 is a selective inhibitor of the Ataxia telangiectasia (A-T) mutated (ATM) protein with an IC50 value of 6.3 nM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5115