Learn More
Sklar Instruments Kuehne Cover Glass Forceps, Sklar, 4in., Square End, Angled, Smooth, Thumb, Stainless Steel. Sklar's Kuehne Cover Glass EA/1.
SDP
Supplier:  Sklar Instruments 193400
Encompass_Preferred
Kuehne Cover Glass Forceps, Sklar, 4 in., Square End, Angled, Smooth, Thumb, Stainless Steel. Sklar's Kuehne Cover Glass Forceps is a thumb instrument used for handling slides for a microscope. It is angled for better support with smooth, square ends.
Catalog No. 50-143-7443