Learn More
Cayman Chemical Kukoamn B 10mg
SDP
Supplier:  Cayman Chemical 2109110 MG
Encompass_Preferred
A natural alkaloid that inhibits LPS and CpG DNA (Kds = 1.24 and 0.66 µM), which are two PAMPs involved in triggering sepsis
Catalog No. 50-298-4984