Learn More
TARGETMOL CHEMICALS INC KURARINONE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5S09935MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Kurarinone a flavanoid derived from shrub Sophora flavescensKurarinone exhibits anti-tumor estrogenic and anti-inflammatory activities it also shows strong inhibitory effect on immune responses. Kurarinone may ameliorate chronic inflammatory skin diseases through the suppression of pathogenic CD4(+) T-cell differentiation and the overall immune response. purity: 99%
Catalog No. 50-257-6644