Learn More
Medchemexpress LLC KVA-D-88 | 2410550-31-9 | 99.8% | 382.19 | C17H11Cl2F2N3O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1391785MG
Encompass_Preferred
KVA-D-88 is a small-molecule, selective phosphodiesterase 4B (PDE4B) inhibitor supplied for research use in pharmacology and neuroscience studies. It has been used in vivo to investigate modulation of reward-related behaviors and cAMP signaling pathways.
  • Selective inhibition of PDE4B, useful for target-specific studies.
  • High chemical purity suitable for research assays.
  • Powder form facilitates formulation and dosing flexibility.
  • Stable under recommended low-temperature storage conditions.
  • Reported molecular weight and formula available for analytical confirmation.
Catalog No. 50-002-22074