Learn More
Selleck Chemical LLC KW-2449 10mg 1000669-72-6
SDP
Supplier:  Selleck Chemical LLC S215810MG
Encompass_Preferred
KW-2449 is a multiple-targeted inhibitor, mostly for Flt3 with IC50 of 6.6 nM, modestly potent to FGFR1, Bcr-Abl and Aurora A; little effect on PDGFR?, IGF-1R, EGFR. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-757-8