Learn More
Medchemexpress LLC HY-13468 5mg Medchemexpress, KW-2478 CAS:819812-04-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134685MG
Encompass_Preferred
Medchemexpress, HY-13468 5mg KW-2478 CAS:819812-04-9 KW-2478 is an inhibitor of Hsp90α, with an IC50 of 3.8 nM, and has antitumor activity against various human hematological tumor cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1374