Learn More
Medchemexpress LLC KW-2478 | 819812-04-9 | >98.0% | C30H42N2O9 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1346810MG
Encompass_Preferred
A potent small-molecule inhibitor of heat shock protein 90 (Hsp90) with a reported IC50 of approximately 3.8 nM. It demonstrates antitumor activity in hematological cancer models and has been evaluated in early-phase clinical studies for B-cell malignancies. Provided as a crystalline solid for research use.
  • Potent Hsp90 inhibition (IC50 ≈ 3.8 nM).
  • Demonstrated antitumor activity against hematological tumor cells.
  • Evaluated in preclinical and early-phase clinical studies.
  • High reported purity suitable for research (≥98%).
  • Supplied as a crystalline solid for laboratory use.
  • Intended for research use only, not for human or veterinary use.
Catalog No. 50-002-59231