Learn More
Supply Solutions Shodex - KW403-4F Aqueous SEC. High performance semi-micro column of PRTEIN KW-800 series. 4.6x300 mm, 3um. Max.Pore 800A.
Supplier:  Supply Solutions SHODEX F6989202
Encompass_Preferred
Shodex - KW403-4F Aqueous SEC. High performance semi-micro column of PRTEIN KW-800 series. 4.6x300 mm, 3um. Max.Pore 800A.
Catalog No. NC1072732