Learn More
Medchemexpress LLC KWCN-41 | 2913223-17-1 | 99.8% | 307.35 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY149258100MG
Encompass_Preferred
KWCN-41 is a selective and efficient inhibitor of RIPK1 kinase with an IC50 value of 88 nM. It specifically inhibits cell necrosis but does not inhibit apoptosis and also exhibits anti-inflammatory effects.
  • Inhibits L929 cell necroptosis
  • Increases viable cell numbers
  • Inhibits phosphorylation of RIPK1/3 and MLKL
  • Protects HT-29 and U937 cells from necroptosis
  • Improves survival in inflammatory mouse model
Catalog No. 50-003-60865