Learn More
Medchemexpress LLC Kx-01-191 | 2211903-84-1 | 98.21% | 100 MG

Supplier: Medchemexpress LLC HY128721100MG
KX-01-191 (compound 5c′) is identified as a tin-precursor. It is intended for research use only and not for sale to patients.
- Purity: 98.21%
- For research use only
- Molecular weight: 552.34
- Formula: C28H39N3OSn
- Appearance: Solid
- Color: White to off-white
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 6 months, -20°C for 1 month
- Solubility (in vitro): DMSO: ≥ 100 mg/mL (181.05 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.