Learn More
Medchemexpress LLC KX-01-191 | 2211903-84-1 | 99.87% | 25 MG

Supplier: Medchemexpress LLC 25MGHY128721
KX-01-191 (compound 5c′) is a tin-precursor intended for research use only. It appears as a white to off-white solid.
- Molecular weight: 552.34
- Formula: C28H39N3OSn
- Soluble in DMSO at ≥ 100 mg/mL (181.05 mM) for in vitro use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.