Learn More
Apexbio Technology LLC KX2-391 dihydrochloride 1038395-65-1 10mg
SDP
Supplier:  Apexbio Technology LLC A353510
Encompass_Preferred
KX2-391 dihydrochloride (CAS 1038395-65-1) is an orally bioavailable small molecule that inhibits Src tyrosine kinase through selective interaction with the peptide substrate binding site rather than the ATP-binding site By blocking this non-catalytic domain KX2-391 disrupts Src kinase-mediated signaling pathways involved in tumor cell proliferation and metastatic processes Src kinases are frequently upregulated in various malignancies underscoring their importance in cancer biology KX2-391 is widely utilized in preclinical studies investigating Src-driven mechanisms of oncogenesis and tumor progression
Catalog No. 50-000-93697