Learn More
Medchemexpress LLC KY-556 | 110816-78-9 | 725.57 | 1 ML
Supplier: Medchemexpress LLC HYU0014810MM/1ML
KY-556 is a promising and orally-active pro-agent of disodium cromoglycate (DSCG) against allergic diseases. This solution is for research use only.
- Orally-active pro-agent against allergic diseases
- Provided as a 10 mM solution in 1 mL DMSO
- Ready for reconstitution
- In-stock